Microcentrifuge tubes, SafeSeal
Leverancier: SARSTEDT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
72.706 72.706.200 72.695.001 72.695.002 72.695.004 72.695.005 72.695.006 72.695.007 72.695.200 72.704 72.704.002 72.706.002 72.704.004 72.704.005 72.704.006 72.695.500
SARS72.706EA 0 EUR
SARS72.706 SARS72.706.200 SARS72.695.001 SARS72.695.002 SARS72.695.004 SARS72.695.005 SARS72.695.006 SARS72.695.007 SARS72.695.200 SARS72.704 SARS72.704.002 SARS72.706.002 SARS72.704.004 SARS72.704.005 SARS72.704.006 SARS72.695.500
Microcentrifuge tubes, SafeSeal
Buisjes Microbuisjes
PP, translucent.
  • Wide hinge for easy and precise closure
  • Large writing space on lid, graduated
  • Centrifugation to 30000×g (2,0 ml tubes: to 25000×g)
For transport, storage and sample processing in the laboratory. DNA-/RNase-free tubes and screw caps for the storage of high-grade synthesised oligonucleotides, enzymes, buffers etc. Retaining cams ensure a safe fit also under thermal strain.