Cartridge burner, high temperature
Leverancier: Usbeck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
USBE1424EA 66.2 EUR
USBE1424
Cartridge burner, high temperature
Verwarmers Gasbranders
High temperature burner with piezo electric ignition and needle valve. Maximum temperature of 1600 °C.