Clamps, flexible, with bosshead, swivel type
Leverancier: USBECK Laborgeräte
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
2680 2682
USBE2680EA 50.1 EUR
USBE2680 USBE2682
Clamps, flexible, with bosshead, swivel type
Klemmen Klemmen voor algemeen gebruik
Flexible clamps with bosshead (swivel type). Made of zinc die-cast and brass, nickel plated.
  • With vinyl coated jaws
  • Three-pronged or round jaws
Length: 300 mm