Aspirator bottles, glass, with stopper, PYREX®
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1569/16D 1569/18D 1569/22D
BIBB1569/16DEA 380 EUR
BIBB1569/16D BIBB1569/18D BIBB1569/22D
Aspirator bottles, glass, with stopper, PYREX®
Flessen Doseerflessen
Made of PYREX®, borosilicate glass.
  • Ground side socket (size 29/32) to accept stopcock assemblies
  • Ground neck
  • Supplied with glass stopper
Bottle with grounded side-socket.
Certificaten: Complies with ISO 4796