PP frits for SPE cartridges
Leverancier: VARIAN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
12131019 12131020 12131021 12131022 12131023 12131024
551-0367EA 57 EUR
551-0367 551-0368 551-0369 551-0370 VARI12131023 VARI12131024
PP frits for SPE cartridges
Filters Voorkolom filters en frits
PP frit, are made with high-grade, clean PE for clean extracts.
  • Pre-cut to correct size for accuracy
  • Use with reservoirs or custom packing
These frits are pre-cut to fit into bond elut reservoirs for use in filtration applications or for custom SPE sorbent packing.