Tripod stands
Leverancier: USBECK Laborgeräte
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
2031 2033
USBE2031EA 40.5 EUR
USBE2031 USBE2033
Tripod stands
Staafstatieven
Retort stand base in tripod shape made from powder-coated steel.
  • For rods up to 13 mm Ø
  • With hinged screw M 6
Thickness: 10 mm