Microtubes, prepared
Leverancier: SARSTEDT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
41.1378.005 41.1505.005 41.1503.005 41.1504.005 41.1395.005 41.1500.005 41.1350.005 41.1392.005 41.1393.005 41.1394.005 41.1501.005 41.1506 41.1506.002 41.1506.005
SARS41.1378.005EA 0 EUR
SARS41.1378.005 SARS41.1505.005 SARS41.1503.005 SARS41.1504.005 SARS41.1395.005 SARS41.1500.005 SARS41.1350.005 SARS41.1392.005 SARS41.1393.005 SARS41.1394.005 SARS41.1501.005 SARS41.1506 SARS41.1506.002 SARS41.1506.005
Microtubes, prepared
Buisjes Microbuisjes
PP, translucent with PP screw cap.
  • Type of preparation: Trisodium citrate solution
  • With paper label
  • Graduated
  • Recommended centrifugation: 1800×g
With screw cap or push-on cap.