Vacuum filtration systems, Filtropur V and Filtropur BT
Leverancier: SARSTEDT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
83.1822 83.1822.001 83.1822.100 83.1822.101 83.1823 83.1823.001 83.1823.100 83.1823.101
SARS83.1822EA 0 EUR
SARS83.1822 SARS83.1822.001 SARS83.1822.100 SARS83.1822.101 SARS83.1823 SARS83.1823.001 SARS83.1823.100 SARS83.1823.101
Vacuum filtration systems, Filtropur V and Filtropur BT
Filters Flesfilters
These units are ideal for the cold sterilisation of eg. cell culture media and protein solutions. PES 'high-flow' filter membrane for high throughput and low protein binding.
  • Non pyrogenic, non cytotoxic
  • Fast product identification by indicating the filter material, pore size and lot number on the filter housing
  • Ergonomically shaped receiver bottles
Leveringsinformatie: A sterile screw cap for the collection vessel is included with each filtration unit.
Verpakking: Individually wrapped.