Cryoboxes
Leverancier: SARSTEDT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
93.873.281 93.873.481 93.873.681 93.874.610
SARS93.873.281EA 51.78 EUR
SARS93.873.281 SARS93.873.481 SARS93.873.681 SARS93.874.610
Cryoboxes
Dozen Dozen voor cryobuisjes
Cryoboxes, made of high quality durable polycarbonate.
  • Numerical coding for each tubes enables quick sample identification
  • Optically clear lid and coloured base with large apertures for swift ventilation
  • Autoclavable
For storage of tubes at deep freeze range to −196 °C.