Transport Boxes
Leverancier: SARSTEDT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
95.1715 95.1717 95.1717.003 95.1717.011
SARS95.1715EA 0 EUR
SARS95.1715 SARS95.1717 SARS95.1717.003 SARS95.1717.011
Transport Boxes
Dozen Transportdozen
Transport case housing is manufactured from durable polyethylene and offers optimum protection for primary receptacles during transport.
  • Economical, flexible
  • Central one-hand closure
  • Lateral tabs for easy attachment of straps or restraints
  • Transparent window label for easy identification
Transport cases are suitable for transportation of diagnostic samples and blood products.
Certificaten: Complies with the requirement of P650 packaging instructions for class UN3373 of the ADR and RID.