Spare PTFE Keys for Stopcocks, PYREX®
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SST303K SST103K SST304K SST203K SST102K SST104K
BIBBSST303KEA 295 EUR
BIBBSST303K BIBBSST103K BIBBSST304K BIBBSST203K BIBBSST102K BIBBSST104K
Spare PTFE Keys for Stopcocks, PYREX®
Afsluitersleutels
Fully interchangeable PTFE keys for Pyrex® stopcocks.
  • Supplied complete with plastic screw thread retaining device
  • Self lubricating, avoiding the need for stopcock grease
  • Made to a taper of 1:10 giving a combination of performance and ease of operation
Leveringsinformatie: Key is supplied complete with plastic screw thread retaining device.