Faeces containers
Leverancier: SARSTEDT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
80.734.301 80.620 80.621 80.622 80.622.111 80.623 80.623.111 80.734 80.734.001 80.734.311 80.734.401
SARS80.734.301EA 0 EUR
SARS80.734.301 SARS80.620 SARS80.621 SARS80.622 SARS80.622.111 SARS80.623 SARS80.623.111 SARS80.734 SARS80.734.001 SARS80.734.311 SARS80.734.401
Faeces containers
Diagnostische systemen Clinical Sample Collection/Preparation Faeces Sample Collection
Containers with sampling scoops enables clean and easy collection of faeces samples.
  • To collect defined faeces volume of 1 ml, approximately 1 g
  • Sterile or non sterile
  • Labelled or opaque options available