Urine collection container, for snap-on lid
Leverancier: SARSTEDT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
75.565 76.566
SARS75.565EA 8.33 EUR
SARS75.565 SARS76.566
Urine collection container, for snap-on lid
Diagnostische systemen Clinical Sample Collection/Preparation Urine Sample Collection/Preparation
PP, translucent.
  • Graduated
  • Excellent chemical resistance
  • Excellent temperature resistance
Disposable neutral container. For collection and storage of urine.
Bestelinformatie: Snap-on lids must be ordered seperately.
Opgelet: *De product(en) op deze pagina vallen buiten het toepassingsgebied van de EU-richtlijn 2019/904