Urine collection container, with snap-on lid
Leverancier: SARSTEDT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
75.1355 75.1355.100 76.1414 76.1414.100 76.1477 76.1477.100
SARS75.1355EA 59.8 EUR
SARS75.1355 SARS75.1355.100 SARS76.1414 SARS76.1414.100 SARS76.1477 SARS76.1477.100
Urine collection container, with snap-on lid
Diagnostische systemen Clinical Sample Collection/Preparation Urine Sample Collection/Preparation
PP, highly transparent.
  • Graduated
  • Excellent chemical resiatnce
  • Excellent temperature resistance
Disposable container for collection and storage of urine.
Bestelinformatie: Snap-on lid must be ordered separately.
Opgelet: *De product(en) op deze pagina vallen buiten het toepassingsgebied van de EU-richtlijn 2019/904