Bottle-top dispenser, Calibrex™
Leverancier: Socorex Isba
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
520.002 520.005 520.010
613-4855EA 493 EUR
613-4855 613-4856 613-4857
Bottle-top dispenser, Calibrex™
Flesdispensers
With fix or adjustable volume.
  • PFA coated plungerwith broad chemical resistance
  • QR coded chemical compatibility
  • Integrated calibration key
  • Quick disassembling, no tool required
  • Autoclavable at 121 °C fully assembled
This model is ranging 1 to 10 ml. The Calibrex™ combine a clever simple construction and a universal chemical compatibility. Appreciated for its smooth, effort-less activation and sturdiness, the base thread fits 32 mm bottle neck diameter.
Leveringsinformatie: Supplied with three PP adapters for 28, 40 and 45 mm bottle neck diameters - See bottle-neck adapters for additional sizes and materials - and a tube.