Micro filter funnels
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
3780/02M 3780/04M
BIBB3780/02MEA 618 EUR
BIBB3780/02M BIBB3780/04M
Micro filter funnels
Trechters Filtertrechters Microfiltertrechters
Micro funnels, with sintered disc, made of PYREX® borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Ideal for filtering small amounts of material.