Cell culture flask with quick release cap
Leverancier: SARSTEDT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
83.3910.502 83.3912.502 83.1813.502 83.1810.500 83.1812.500 83.1813.500
SARS83.3910.502EA 0 EUR
SARS83.3910.502 SARS83.3912.502 SARS83.1813.502 SARS83.1810.500 SARS83.1812.500 SARS83.1813.500
Cell culture flask with quick release cap
Kolven Celcultuurflessen
Cell culture flask, PS, clear, with PE cap, hydrophobic growth surfaces for suspension and hybridoma cells.
  • Pyrogen-free
  • Non-cytotoxic
  • Canted neck