Filter tubes, plain neck
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
2110/06M 2110/08M
BIBB2110/06MEA 888 EUR
BIBB2110/06M BIBB2110/08M
Filter tubes, plain neck
Filters Filterbuizen
These filter tubes can be used for small scale filtrations. They are ideal for use with Pyrex® short stem funnels.