Anti-CCL25 Rabbit Monoclonal Antibody [clone: EPR21835-220]

Leverancier: Abcam
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AB259495-100 AB259495-1
ABCAAB259495-100EA 938 EUR
ABCAAB259495-100 ABCAAB259495-1
Anti-CCL25 Rabbit Monoclonal Antibody [clone: EPR21835-220]
Antilichamen
Rabbit monoclonal [EPR21835-220] to CCL25 - BSA and Azide free (Capture).

Type: Primary
Antigen: CCL25
Clonality: Monoclonal
Clone: EPR21835-220
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR