Anti-Brachyury/Bry Rabbit Monoclonal Antibody [clone: 54]
Leverancier: Abcam
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AB259260-100 AB259260-1MG AB259260-20UG
ABCAAB259260-100EA 713 EUR
ABCAAB259260-100 ABCAAB259260-1MG ABCAAB259260-20UG
Anti-Brachyury/Bry Rabbit Monoclonal Antibody [clone: 54]
Antilichamen
Rabbit monoclonal [54] to Brachyury/Bry.
Opgelet: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Brachyury/Bry
Clonality: Monoclonal
Clone: 54
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human