Anti-gamma Catenin Mouse Monoclonal Antibody [clone: 15/γ-Catenin]

Leverancier: Abcam
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AB305241-10 AB305241-100 AB305241-20UG AB305241-1MG
ABCAAB305241-10EA 219 EUR
ABCAAB305241-10 ABCAAB305241-100 ABCAAB305241-20UG ABCAAB305241-1MG
Anti-gamma Catenin Mouse Monoclonal Antibody [clone: 15/γ-Catenin]
Antilichamen
Mouse monoclonal [15/gamma-Catenin] to gamma Catenin.

Opgelet: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: gamma Catenin
Clonality: Monoclonal
Clone: 15/γ-Catenin
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human; Mouse; Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR