Anti-Delta Opioid Receptor Rabbit Monoclonal Antibody [clone: EPR24378-193]

Leverancier: Abcam
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AB283692-100 AB283692-20UL AB283692-1ML
ABCAAB283692-100EA 713 EUR
ABCAAB283692-100 ABCAAB283692-20UL ABCAAB283692-1ML
Anti-Delta Opioid Receptor Rabbit Monoclonal Antibody [clone: EPR24378-193]
Antilichamen
Rabbit monoclonal [EPR24378-193] to Delta Opioid Receptor.

Opgelet: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Delta Opioid Receptor
Clonality: Monoclonal
Clone: EPR24378-193
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR