Anti-GM-CSF Rabbit Monoclonal Antibody [clone: EPR23689-17]
Leverancier: Abcam
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AB227031-100 AB227031-1ML
ABCAAB227031-100EA 713 EUR
ABCAAB227031-100 ABCAAB227031-1ML
Anti-GM-CSF Rabbit Monoclonal Antibody [clone: EPR23689-17]
Antilichamen
Rabbit monoclonal [EPR23689-17] to GM-CSF.
Opgelet: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: GM-CSF
Clonality: Monoclonal
Clone: EPR23689-17
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human; Mouse