Anti-Integrin alpha V Mouse Monoclonal Antibody [clone: 21 CD5]
Leverancier: Abcam
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AB302640-100 AB302640-10 AB302640-20UG AB302640-1MG
ABCAAB302640-100EA 707 EUR
ABCAAB302640-100 ABCAAB302640-10 ABCAAB302640-20UG ABCAAB302640-1MG
Anti-Integrin alpha V Mouse Monoclonal Antibody [clone: 21 CD5]
Antilichamen
Mouse monoclonal [21/CD51] to Integrin alpha V.
Opgelet: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Integrin alpha V
Clonality: Monoclonal
Clone: 21 CD5
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human; Mouse; Rat