Anti-ZO1 tight junction protein Rabbit Monoclonal Antibody [clone: EPR19945-296]

Leverancier: Abcam
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AB251568-100 AB251568-1
ABCAAB251568-100EA 1020 EUR
ABCAAB251568-100 ABCAAB251568-1
Anti-ZO1 tight junction protein Rabbit Monoclonal Antibody [clone: EPR19945-296]
Antilichamen
Rabbit monoclonal [EPR19945-296] to ZO1 tight junction protein - BSA and Azide free.

Opgelet: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: ZO1 tight junction protein
Clonality: Monoclonal
Clone: EPR19945-296
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR