Anti-DR5 Rabbit Monoclonal Antibody [clone: EPR22276]
Leverancier: Abcam
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AB243847-100 AB243847-1
ABCAAB243847-100EA 1020 EUR
ABCAAB243847-100 ABCAAB243847-1
Anti-DR5 Rabbit Monoclonal Antibody [clone: EPR22276]
Antilichamen
Rabbit monoclonal [EPR22276] to DR5 - BSA and Azide free.
Opgelet: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: DR5
Clonality: Monoclonal
Clone: EPR22276
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human