Anti-PCSK2 Rabbit Monoclonal Antibody [clone: EPR23578-19]

Leverancier: Abcam
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AB274418-100 AB274418-20UL AB274418-1ML
ABCAAB274418-100EA 719 EUR
ABCAAB274418-100 ABCAAB274418-20UL ABCAAB274418-1ML
Anti-PCSK2 Rabbit Monoclonal Antibody [clone: EPR23578-19]
Antilichamen
Rabbit monoclonal [EPR23578-19] to PCSK2.

Opgelet: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: PCSK2
Clonality: Monoclonal
Clone: EPR23578-19
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR