Anti-Carbonic Anhydrase 12/CA12 Rabbit Monoclonal Antibody [clone: EPR14861]
Leverancier: Abcam
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AB236138-100 AB236138-1
ABCAAB236138-100EA 1020 EUR
ABCAAB236138-100 ABCAAB236138-1
Anti-Carbonic Anhydrase 12/CA12 Rabbit Monoclonal Antibody [clone: EPR14861]
Antilichamen
Rabbit monoclonal [EPR14861] to Carbonic Anhydrase 12/CA12 - BSA and Azide free.
Opgelet: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: Carbonic Anhydrase 12/CA12
Clonality: Monoclonal
Clone: EPR14861
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human