Anti-TEMT Rabbit Monoclonal Antibody [clone: EPR13240-105]

Leverancier: Abcam
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AB184573-100 AB184573-1ML
ABCAAB184573-100EA 700 EUR
ABCAAB184573-100 ABCAAB184573-1ML
Anti-TEMT Rabbit Monoclonal Antibody [clone: EPR13240-105]
Antilichamen
Rabbit monoclonal [EPR13240-105] to TEMT.

Opgelet: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: TEMT
Clonality: Monoclonal
Clone: EPR13240-105
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR