Anti-Calcium-independent Phospholipase A2/PLA2G6 Rabbit Monoclonal Antibody [clone: EPR23994-103]
Leverancier: Abcam
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AB278086-1 AB278086-100
ABCAAB278086-1EA 9160 EUR
ABCAAB278086-1 ABCAAB278086-100
Anti-Calcium-independent Phospholipase A2/PLA2G6 Rabbit Monoclonal Antibody [clone: EPR23994-103]
Antilichamen
Rabbit monoclonal [EPR23994-103] to Calcium-independent Phospholipase A2/PLA2G6 - BSA and Azide free.
Opgelet: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Calcium-independent Phospholipase A2/PLA2G6
Clonality: Monoclonal
Clone: EPR23994-103
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Rat; Human