Anti-Histone H3 (acetyl K4) Rabbit Monoclonal Antibody [clone: EPR16596]

Leverancier: Abcam
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AB240195-1 AB240195-100
ABCAAB240195-1EA 9160 EUR
ABCAAB240195-1 ABCAAB240195-100
Anti-Histone H3 (acetyl K4) Rabbit Monoclonal Antibody [clone: EPR16596]
Antilichamen
Rabbit monoclonal [EPR16596] to Histone H3 (acetyl K4) - BSA and Azide free.

Opgelet: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Histone H3
Clonality: Monoclonal
Clone: EPR16596
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR