Anti-Cyclin D3/CCND3 Rabbit Monoclonal Antibody [clone: SP207]

Leverancier: Abcam
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AB183338-100 AB183338-1
ABCAAB183338-100EA 744 EUR
ABCAAB183338-100 ABCAAB183338-1
Anti-Cyclin D3/CCND3 Rabbit Monoclonal Antibody [clone: SP207]
Antilichamen
Rabbit monoclonal [SP207] to Cyclin D3/CCND3.

Opgelet: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Cyclin D3/CCND3
Clonality: Monoclonal
Clone: SP207
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR