Anti-CEBP Delta/CEBPD Rabbit Monoclonal Antibody [clone: EPR23518-259]
Leverancier: Abcam
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AB270410-100 AB270410-1
ABCAAB270410-100EA 1020 EUR
ABCAAB270410-100 ABCAAB270410-1
Anti-CEBP Delta/CEBPD Rabbit Monoclonal Antibody [clone: EPR23518-259]
Antilichamen
Rabbit monoclonal [EPR23518-259] to CEBP Delta/CEBPD - BSA and Azide free.
Opgelet: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: CEBP Delta/CEBPD
Clonality: Monoclonal
Clone: EPR23518-259
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat