Anti-Alcam Rabbit Monoclonal Antibody [clone: EPR22341-191]

Leverancier: Abcam
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AB272281-1 AB272281-100
ABCAAB272281-1EA 8680 EUR
ABCAAB272281-1 ABCAAB272281-100
Anti-Alcam Rabbit Monoclonal Antibody [clone: EPR22341-191]
Antilichamen
Rabbit monoclonal [EPR22341-191] to Alcam - BSA and Azide free (Capture).

Opgelet: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Alcam
Clonality: Monoclonal
Clone: EPR22341-191
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR