Anti-Histone H3 (acetyl K4 + K9 + K14 + K18 + K23 + K27) Rabbit Recombinant Multiclonal Antibody [clone: RM1045]

Leverancier: Abcam
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AB300642-100 AB300642-1
ABCAAB300642-100EA 1020 EUR
ABCAAB300642-100 ABCAAB300642-1
Anti-Histone H3 (acetyl K4 + K9 + K14 + K18 + K23 + K27) Rabbit Recombinant Multiclonal Antibody [clone: RM1045]
Antilichamen
Rabbit recombinant multiclonal [RM1045] to Histone H3 (acetyl K4 + K9 + K14 + K18 + K23 + K27) - BSA and Azide free.

Opgelet: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Histone H3
Clonality: Recombinant Multiclonal
Clone: RM1045
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR