Anti-Cyclophilin B Rabbit Monoclonal Antibody [clone: EPR12703(B)]

Leverancier: Abcam
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AB178397-100 AB178397-250 AB178397-1ML
ABCAAB178397-100EA 475 EUR
ABCAAB178397-100 ABCAAB178397-250 ABCAAB178397-1ML
Anti-Cyclophilin B Rabbit Monoclonal Antibody [clone: EPR12703(B)]
Antilichamen
Rabbit monoclonal [EPR12703(B)] to Cyclophilin B - Loading Control.

Opgelet: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Control
Antigen: Cyclophilin B
Clonality: Monoclonal
Clone: EPR12703(B)
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR