Anti-Kv4.3/KCND3 Mouse Monoclonal Antibody [clone: K75/41]

Leverancier: Abcam
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AB252538-100 AB252538-1MG
ABCAAB252538-100EA 763 EUR
ABCAAB252538-100 ABCAAB252538-1MG
Anti-Kv4.3/KCND3 Mouse Monoclonal Antibody [clone: K75/41]
Antilichamen
Mouse monoclonal [K75/41] to Kv4.3/KCND3.

Opgelet: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Kv4.3/KCND3
Clonality: Monoclonal
Clone: K75/41
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR