Anti-Sodium Potassium ATPase Rat Monoclonal Antibody [clone: EP1845Y]
Leverancier: Abcam
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AB283341-100 AB283341-1
ABCAAB283341-100EA 1020 EUR
ABCAAB283341-100 ABCAAB283341-1
Anti-Sodium Potassium ATPase Rat Monoclonal Antibody [clone: EP1845Y]
Antilichamen
Rat monoclonal [EP1845Y] to Sodium Potassium ATPase - Chimeric – BSA and Azide free.
Opgelet: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Sodium Potassium ATPase
Clonality: Monoclonal
Clone: EP1845Y
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human;Mouse;Rat