Anti-RILPL1 Mouse Monoclonal Antibody [clone: MJF-R41-21]
Leverancier: Abcam
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AB307294-10 AB307294-100 AB307294-20UL AB307294-1ML
ABCAAB307294-10EA 119 EUR
ABCAAB307294-10 ABCAAB307294-100 ABCAAB307294-20UL ABCAAB307294-1ML
Anti-RILPL1 Mouse Monoclonal Antibody [clone: MJF-R41-21]
Antilichamen
Mouse monoclonal [MJF-R41-21] to RILPL1 - Chimeric.
Opgelet: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: RILPL1
Clonality: Monoclonal
Clone: MJF-R41-21
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human;Mouse;Rat