Anti-NKG2D Rabbit Monoclonal Antibody [clone: EPR24072-342] (Alexa Fluor® 647)

Leverancier: Abcam
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AB312904-100UL AB312904-10UL
ABCAAB312904-100ULEA 819 EUR
ABCAAB312904-100UL ABCAAB312904-10UL
Anti-NKG2D Rabbit Monoclonal Antibody [clone: EPR24072-342] (Alexa Fluor® 647)
Antilichamen
Alexa Fluor® 647 Rabbit monoclonal [EPR24072-342] to NKG2D.

Opgelet: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: NKG2D
Clonality: Monoclonal
Clone: EPR24072-342
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR