Anti-TRIM33 Rabbit Monoclonal Antibody [clone: EPR25102-19]

Leverancier: Abcam
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AB300146-100 AB300146-20UL AB300146-1ML
ABCAAB300146-100EA 707 EUR
ABCAAB300146-100 ABCAAB300146-20UL ABCAAB300146-1ML
Anti-TRIM33 Rabbit Monoclonal Antibody [clone: EPR25102-19]
Antilichamen
Rabbit monoclonal [EPR25102-19] to TRIM33.

Opgelet: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: TRIM33
Clonality: Monoclonal
Clone: EPR25102-19
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR