Anti-CSF-1-R Rabbit Monoclonal Antibody [clone: EPR20754-46]

Leverancier: Abcam
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AB244712-100 AB244712-1
ABCAAB244712-100EA 938 EUR
ABCAAB244712-100 ABCAAB244712-1
Anti-CSF-1-R Rabbit Monoclonal Antibody [clone: EPR20754-46]
Antilichamen
Rabbit monoclonal [EPR20754-46] to CSF-1-R - BSA and Azide free (Capture).

Type: Primary
Antigen: CSF-1-R
Clonality: Monoclonal
Clone: EPR20754-46
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR