Anti-SARS-CoV2 orf8 Rabbit Monoclonal Antibody [clone: EPR24851-65]
Leverancier: Abcam
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AB283914-100 AB283914-20UL AB283914-1ML
ABCAAB283914-100EA 675 EUR
ABCAAB283914-100 ABCAAB283914-20UL ABCAAB283914-1ML
Anti-SARS-CoV2 orf8 Rabbit Monoclonal Antibody [clone: EPR24851-65]
Antilichamen
Rabbit monoclonal [EPR24851-65] to SARS-CoV2 orf8.
Type: Primary
Antigen: SARS-CoV2 orf8
Clonality: Monoclonal
Clone: EPR24851-65
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now