Anti-PSGL-1 Rabbit Monoclonal Antibody [clone: EPR22504-36] (APC (Allophycocyanin))
Leverancier: Abcam
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AB313947-100UL AB313947-10UL
ABCAAB313947-100ULEA 819 EUR
ABCAAB313947-100UL ABCAAB313947-10UL
Anti-PSGL-1 Rabbit Monoclonal Antibody [clone: EPR22504-36] (APC (Allophycocyanin))
Antilichamen
APC Rabbit monoclonal [EPR22504-36] to PSGL-1.
Opgelet: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: PSGL-1
Clonality: Monoclonal
Clone: EPR22504-36
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human