Anti-Histone H3 (acetyl K27) Mouse Monoclonal Antibody [clone: EPR20551-225]

Leverancier: Abcam
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AB317801-100UG AB317801-1MG
ABCAAB317801-100UGEA 1020 EUR
ABCAAB317801-100UG ABCAAB317801-1MG
Anti-Histone H3 (acetyl K27) Mouse Monoclonal Antibody [clone: EPR20551-225]
Antilichamen
Mouse monoclonal [EPR20551-225] to Histone H3 (tri methyl K4) - Chimeric – BSA and Azide free.

Opgelet: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Histone H3
Clonality: Monoclonal
Clone: EPR20551-225
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human;Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR