Anti-LY75/DEC-205 Rabbit Monoclonal Antibody [clone: EPR27222-93] (Alexa Fluor® 647)

Leverancier: Abcam
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AB320783-10UL AB320783-100UL
ABCAAB320783-10ULEA 238 EUR
ABCAAB320783-10UL ABCAAB320783-100UL
Anti-LY75/DEC-205 Rabbit Monoclonal Antibody [clone: EPR27222-93] (Alexa Fluor® 647)
Antilichamen
Alexa Fluor® 647 Rabbit monoclonal [EPR27222-93] to LY75/DEC-205.

Opgelet: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: LY75/DEC-205
Clonality: Monoclonal
Clone: EPR27222-93
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR