Anti-LIGHT/TNFSF14 Rabbit Monoclonal Antibody [clone: EPR23428-536] (Alexa Fluor® 488)

Leverancier: Abcam
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AB319123-10UL AB319123-100UL
ABCAAB319123-10ULEA 0 EUR
ABCAAB319123-10UL ABCAAB319123-100UL
Anti-LIGHT/TNFSF14 Rabbit Monoclonal Antibody [clone: EPR23428-536] (Alexa Fluor® 488)
Antilichamen
Alexa Fluor® 488 Rabbit monoclonal [EPR23428-536] to LIGHT/TNFSF14.

Opgelet: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: LIGHT/TNFSF14
Clonality: Monoclonal
Clone: EPR23428-536
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR