Anti-Plexin B1 Rabbit Monoclonal Antibody [clone: EPR26309-65]

Leverancier: Abcam
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AB316313-1MG AB316313-100UG
ABCAAB316313-1MGEA 9160 EUR
ABCAAB316313-1MG ABCAAB316313-100UG
Anti-Plexin B1 Rabbit Monoclonal Antibody [clone: EPR26309-65]
Antilichamen
Rabbit monoclonal [EPR26309-65] to Plexin B1 - BSA and Azide free.

Opgelet: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Plexin B1
Clonality: Monoclonal
Clone: EPR26309-65
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR