Anti-CCR9 Rabbit Recombinant Multiclonal Antibody [clone: RM2031]
Leverancier: Abcam
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AB315485-1MG AB315485-100UG
ABCAAB315485-1MGEA 9160 EUR
ABCAAB315485-1MG ABCAAB315485-100UG
Anti-CCR9 Rabbit Recombinant Multiclonal Antibody [clone: RM2031]
Antilichamen
Rabbit recombinant multiclonal [RM2031] to CCR9 - BSA and Azide free.
Opgelet: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: CCR9
Clonality: Recombinant Multiclonal
Clone: RM2031
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat