Anti-Bcl6 Rabbit Recombinant Multiclonal Antibody [clone: RM1183]

Leverancier: Abcam
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AB317338-100UG AB317338-1MG
ABCAAB317338-100UGEA 1020 EUR
ABCAAB317338-100UG ABCAAB317338-1MG
Anti-Bcl6 Rabbit Recombinant Multiclonal Antibody [clone: RM1183]
Antilichamen
Rabbit recombinant multiclonal [RM1183] to Bcl6 - BSA and Azide free.

Opgelet: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Bcl6
Clonality: Recombinant Multiclonal
Clone: RM1183
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR